Thermo Scientific Chemicals

2-Methyl-3-butyn-2-ol, 98%, Thermo Scientific Chemicals

Catalog number: L07593.0F
2500 mL, Each
Thermo Scientific Chemicals

2-Methyl-3-butyn-2-ol, 98%, Thermo Scientific Chemicals

Catalog number: L07593.0F
2500 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L07593.0F
also known as L07593-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
115-19-5
IUPAC Name
2-methylbut-3-yn-2-ol
Molecular Formula
C5H8O
InChI Key
CEBKHWWANWSNTI-UHFFFAOYSA-N
SMILES
CC(C)(O)C#C
Appearance (Color)
Clear, colourless
Assay (GC)
>97.5%
Refractive Index
1.4190 - 1.4230 @20?C
Form
liquid

Description

2-Methyl-3-butyn-2-ol is used as an intermediate in the manufacture of products for the agrochemical and specialty chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-3-butyn-2-ol is used as an intermediate in the manufacture of products for the agrochemical and specialty chemical industry.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text