2-Methyl-3-butyn-2-ol, 98%, Thermo Scientific Chemicals
2-Methyl-3-butyn-2-ol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-3-butyn-2-ol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L07593.AE
Price (USD)/ Each
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS115-19-5
IUPAC Name2-methylbut-3-yn-2-ol
Molecular FormulaC5H8O
InChI KeyCEBKHWWANWSNTI-UHFFFAOYSA-N
SMILESCC(C)(O)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colourless
Assay (GC)>97.5%
Refractive Index1.4190 - 1.4230 @20?C
Formliquid
2-Methyl-3-butyn-2-ol is used as an intermediate in the manufacture of products for the agrochemical and specialty chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-3-butyn-2-ol is used as an intermediate in the manufacture of products for the agrochemical and specialty chemical industry.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Edward T. Sabourin.; Anatoli Onopchenko. A convenient synthesis of 4-ethynylphthalic anhydride via 2-methyl-3-butyn-2-ol. J. Org. Chem. 1983, 48, (25), 5135-5137.
  2. D. Boyall.; F. López.; H. Sasaki.; D. Frantz.; E. M. Carreira. Enantioselective Addition of 2-Methyl-3-butyn-2-ol to Aldehydes: Preparation of 3-Hydroxy-1-butynes. Org. Lett. 2000, 2, (26), 4233-4236.
  3. Adduct of acetylene and acetone which acts as an acetylene equivalent in the Pd-catalyzed (Sonogashira) coupling reaction with aryl or heteroaryl halides, permitting a one-pot diarylation by different halides: Synthesis, 571 (1984):
  4. Alternatively, the protecting group can be removed from the intermediate by base to give monoarylacetylenes: J. Org. Chem., 48, 5135 (1983); 57, 6998 (1992). For an improved procedure, see: Synthesis, 589 (1996).