Thermo Scientific Chemicals

Di-n-butyl sulfide, 98%, Thermo Scientific Chemicals

Catalog number: L07599.AE
100 mL, Each
Thermo Scientific Chemicals

Di-n-butyl sulfide, 98%, Thermo Scientific Chemicals

Catalog number: L07599.AE
100 mL, Each
Quantity
Catalog number: L07599.AE
also known as L07599-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
544-40-1
IUPAC Name
1-(butylsulfanyl)butane
Molecular Formula
C8H18S
InChI Key
HTIRHQRTDBPHNZ-UHFFFAOYSA-N
SMILES
CCCCSCCCC
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Refractive Index
1.4510-1.4550 @ 20?C

Description

Di-n-butyl sulfide is generally used as an agricultural intermediate, a sulfiding agent, a refinery catalyst, a lubricant additive, a gas odorant and a processing aid in mining applications. It finds application as an internal standard in X-ray fluorescence spectrometry (XRF) measurements of sulfur in oils and other liquid hydrocarbon matrices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-butyl sulfide is generally used as an agricultural intermediate, a sulfiding agent, a refinery catalyst, a lubricant additive, a gas odorant and a processing aid in mining applications. It finds application as an internal standard in X-ray fluorescence spectrometry (XRF) measurements of sulfur in oils and other liquid hydrocarbon matrices.

Solubility
Silghtly miscible with water. Miscible with olive oil and almond oil.

Notes
Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text