Thermo Scientific Chemicals

alpha-Angelicalactone, 98%, Thermo Scientific Chemicals

Catalog number: L07602.22
100 g, Each
Thermo Scientific Chemicals

alpha-Angelicalactone, 98%, Thermo Scientific Chemicals

Catalog number: L07602.22
100 g, Each
Quantity
Catalog number: L07602.22
also known as L07602-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
591-12-8
IUPAC Name
5-methyl-2,3-dihydrofuran-2-one
Molecular Formula
C5H6O2
InChI Key
QOTQFLOTGBBMEX-UHFFFAOYSA-N
SMILES
CC1=CCC(=O)O1
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Refractive Index
1.4460-1.4500 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms

Description

alpha-Angelicalactone acts as acancer chemopreventive agent. It is used in the synthesis of glutathione and the activity of glutathione-S-transferase and UDP-glucunonosyltransferase detoxification enzymes in esophagus, stomach, intestine, and liver. It is used in oral care formulations to round out mint notes and in coffee flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-Angelicalactone acts as acancer chemopreventive agent. It is used in the synthesis of glutathione and the activity of glutathione-S-transferase and UDP-glucunonosyltransferase detoxification enzymes in esophagus, stomach, intestine, and liver. It is used in oral care formulations to round out mint notes and in coffee flavors.

Solubility
Soluble in water (5g/100mL) and alcohol.

Notes
Moisture-sensitive. Store under inert gas. Protect from moisture. Incompatible materials are Oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text