Thermo Scientific Chemicals

1,2-Epoxyhexane, 96%, Thermo Scientific Chemicals

Catalog number: L07609.22
100 g, Each
Thermo Scientific Chemicals

1,2-Epoxyhexane, 96%, Thermo Scientific Chemicals

Catalog number: L07609.22
100 g, Each
Quantity
Catalog number: L07609.22
also known as L07609-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1436-34-6
IUPAC Name
2-butyloxirane
Molecular Formula
C6H12O
InChI Key
WHNBDXQTMPYBAT-UHFFFAOYNA-N
SMILES
CCCCC1CO1
Assay (GC)
≥95.0%
Refractive Index
1.4035-1.4095 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid
Identification (FTIR)
Conforms

Description

1,2-Epoxyhexane is used as chemical reagent, organic intermediates, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxyhexane is used as chemical reagent, organic intermediates, fine chemicals, pharmaceutical research and development.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text