Thermo Scientific Chemicals

1-Bromopentadecane, 97%, Thermo Scientific Chemicals

Catalog number: L07618.18
50 g, Each
Thermo Scientific Chemicals

1-Bromopentadecane, 97%, Thermo Scientific Chemicals

Catalog number: L07618.18
50 g, Each
Quantity
Catalog number: L07618.18
also known as L07618-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
629-72-1
IUPAC Name
1-bromopentadecane
Molecular Formula
C15H31Br
InChI Key
JKOTZBXSNOGCIF-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCBr
Form
Crystals or powder or crystalline powder or clear liquid as melt
Appearance (Color)
Colorless to white to pale yellow
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.4590-1.4640 @ 20?C

Description

1-Bromopentadecane has been used as regent in the synthesis of (S)-eicos-(4E)-en-1-yn-3-ol, a bioactive component of the marine sponge Cribrochalina vasculum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopentadecane has been used as regent in the synthesis of (S)-eicos-(4E)-en-1-yn-3-ol, a bioactive component of the marine sponge Cribrochalina vasculum.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text