Thermo Scientific Chemicals

1,2-Epoxyoctadecane, tech. 85%, Thermo Scientific Chemicals

Catalog number: L07622.14
25 g, Each
Thermo Scientific Chemicals

1,2-Epoxyoctadecane, tech. 85%, Thermo Scientific Chemicals

Catalog number: L07622.14
25 g, Each
Quantity
Catalog number: L07622.14
also known as L07622-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7390-81-0
IUPAC Name
2-hexadecyloxirane
Molecular Formula
C18H36O
InChI Key
QBJWYMFTMJFGOL-UHFFFAOYNA-N
SMILES
CCCCCCCCCCCCCCCCC1CO1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or fused solid or lumps
Assay (GC)
≥80.0%

Description

1,2-Epoxyoctadecane can be used to produce octadecane-1,2-diol at heating. It is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxyoctadecane can be used to produce octadecane-1,2-diol at heating. It is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Insoluble in water.

Notes
Protect from moisture. Store away from incompatible materials such as oxidizing agents. Moisture-sensitive. Incompatible with strong acids, peroxides, caustics.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text