Thermo Scientific Chemicals

Ethyl 2-iodobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: L07650.06
5 g, Each
Thermo Scientific Chemicals

Ethyl 2-iodobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: L07650.06
5 g, Each
Quantity
Catalog number: L07650.06
also known as L07650-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1829-28-3
Form
Liquid
Refractive Index
1.5825-1.5865 @ 20°C (non-U.S. specification)
GC/MS Analysis
Conforms (U.S. specification)
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%

Description

Ethyl 2-iodobenzoate is used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-iodobenzoate is used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

Solubility
Difficult to mix.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text