2-Fluoro-DL-phenylalanine, 98%, Thermo Scientific Chemicals
2-Fluoro-DL-phenylalanine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluoro-DL-phenylalanine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L07654.03
also known as L07654-03
Price (USD)/ Each
34.80
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
34.80
Add to cart
2-Fluoro-DL-phenylalanine, 98%, Thermo Scientific Chemicals
Catalog numberL07654.03
Price (USD)/ Each
34.80
-
Add to cart
Chemical Identifiers
CAS2629-55-2
IUPAC Name(2S)-2-azaniumyl-3-(2-fluorophenyl)propanoate
Molecular FormulaC9H10FNO2
InChI KeyNYCRCTMDYITATC-QMMMGPOBSA-N
SMILES[NH3+][C@@H](CC1=CC=CC=C1F)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5% (Dry wt. basis)
Assay (HPLC)≥97.5%
Water Content (Karl Fischer Titration)≤5.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only