1-Hexen-3-one, 90+%, stab. with 0.5% 4-methoxyphenol, Thermo Scientific Chemicals
1-Hexen-3-one, 90+%, stab. with 0.5% 4-methoxyphenol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Hexen-3-one, 90+%, stab. with 0.5% 4-methoxyphenol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L07660.14
also known as L07660-14
Price (USD)/ Each
468.65
Online exclusive
520.00 
Save 51.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
468.65
Online exclusive
520.00 
Save 51.35 (10%)
Add to cart
1-Hexen-3-one, 90+%, stab. with 0.5% 4-methoxyphenol, Thermo Scientific Chemicals
Catalog numberL07660.14
Price (USD)/ Each
468.65
Online exclusive
520.00 
Save 51.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1629-60-3
IUPAC Namehex-1-en-3-one
Molecular FormulaC6H10O
InChI KeyJTHNLKXLWOXOQK-UHFFFAOYSA-N
SMILESCCCC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Solution Test10% w/v in diethyl ether: clear solution
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥90.0%
View more
1-Hexen-3-one is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexen-3-one is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Soluble in water (slightly).

Notes
Store at 4°C. Protect from heat. Incompatible with oxidizing agents, heat, light.
RUO – Research Use Only