Chloroacetaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals
Chloroacetaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloroacetaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number L07661.22
also known as L07661-22
Price (USD)/ Each
48.90
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
48.90
Add to cart
Chloroacetaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals
Catalog numberL07661.22
Price (USD)/ Each
48.90
-
Add to cart
Chemical Identifiers
CAS621-62-5
IUPAC Name2-chloro-1,1-diethoxyethane
Molecular FormulaC6H13ClO2
InChI KeyOVXJWSYBABKZMD-UHFFFAOYSA-N
SMILESCCOC(CCl)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Formliquid
Refractive Index1.4140-1.4180
Appearance (Color)Clear, colourless to pale yellow
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only