Ethyl acetamidocyanoacetate, 98%, Thermo Scientific Chemicals
Ethyl acetamidocyanoacetate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl acetamidocyanoacetate, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
10 g
Catalog number L07674.09
also known as L07674-09
Price (USD)/ Each
73.50
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
73.50
Add to cart
Ethyl acetamidocyanoacetate, 98%, Thermo Scientific Chemicals
Catalog numberL07674.09
Price (USD)/ Each
73.50
-
Add to cart
Chemical Identifiers
CAS4977-62-2
IUPAC Nameethyl 2-cyano-2-acetamidoacetate
Molecular FormulaC7H10N2O3
InChI KeySLIRLABNGAZSHX-UHFFFAOYNA-N
SMILESCCOC(=O)C(NC(C)=O)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)125.5-131.5°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only