Thermo Scientific Chemicals

2-Bromo-2'-methoxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: L07696.03
1 g, Each
Thermo Scientific Chemicals

2-Bromo-2'-methoxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog number: L07696.03
1 g, Each
Quantity
Catalog number: L07696.03
also known as L07696-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
31949-21-0
IUPAC Name
2-bromo-1-(2-methoxyphenyl)ethan-1-one
Molecular Formula
C9H9BrO2
InChI Key
GKNCPTLOPRDYMH-UHFFFAOYSA-N
SMILES
COC1=CC=CC=C1C(=O)CBr
Melting Point (clear melt)
41.0-47.0?C
Appearance (Color)
White to cream to brown to gray
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%

Description

2-Bromo-2'-methoxyacetophenone is used as a pharmaceutical intermediate .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-2′-methoxyacetophenone is used as a pharmaceutical intermediate .

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text