2-Bromo-2'-methoxyacetophenone, 98%, Thermo Scientific Chemicals
2-Bromo-2'-methoxyacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-2'-methoxyacetophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number L07696.06
also known as L07696-06
Price (USD)/ Each
122.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
122.00
Add to cart
2-Bromo-2'-methoxyacetophenone, 98%, Thermo Scientific Chemicals
Catalog numberL07696.06
Price (USD)/ Each
122.00
-
Add to cart
Chemical Identifiers
CAS31949-21-0
IUPAC Name2-bromo-1-(2-methoxyphenyl)ethan-1-one
Molecular FormulaC9H9BrO2
InChI KeyGKNCPTLOPRDYMH-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)41.0-47.0?C
Appearance (Color)White to cream to brown to gray
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
2-Bromo-2'-methoxyacetophenone is used as a pharmaceutical intermediate .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-2′-methoxyacetophenone is used as a pharmaceutical intermediate .

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Charles E. Spivak.; Francis L. Harris. Semipinacolic deamination of 2-amino-1-(2-methoxyphenyl)-1-phenylethanol. J. Org. Chem. 1972, 37, (15), 2494-2497.
  2. Yuzo Kawada.; Hiizu Iwamura. Carbonation of 1-triptycyllithium taking place via electron transfer-recombination mechanism. J. Org. Chem. 1981, 46, (16), 3357-3359.