Thermo Scientific Chemicals

D-Lysine monohydrochloride, 98%

Catalog number: L07710.06
5 g, Each
Thermo Scientific Chemicals

D-Lysine monohydrochloride, 98%

Catalog number: L07710.06
5 g, Each
Quantity
Catalog number: L07710.06
also known as L07710-06
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
7274-88-6
IUPAC Name
(2R)-2,6-diaminohexanoic acid hydrochloride
Molecular Formula
C6H15ClN2O2
InChI Key
BVHLGVCQOALMSV-XZNNNFJINA-N
SMILES
Cl.NCCCC[C@@H](N)C(O)=O
Form
Crystalline powder or powder
Optical Rotation
-20.5 ? 1.5? (c=5 in 5N HCl)
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5% (Dry wt. basis)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text