Thermo Scientific Chemicals

Benzoyleneurea, 98%, Thermo Scientific Chemicals

Catalog number: L07763.14
25 g, Each
Thermo Scientific Chemicals

Benzoyleneurea, 98%, Thermo Scientific Chemicals

Catalog number: L07763.14
25 g, Each
Quantity
Catalog number: L07763.14
also known as L07763-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86-96-4
IUPAC Name
1,2,3,4-tetrahydroquinazoline-2,4-dione
Molecular Formula
C8H6N2O2
InChI Key
SDQJTWBNWQABLE-UHFFFAOYSA-N
SMILES
O=C1NC(=O)C2=CC=CC=C2N1
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%
Appearance (Color)
White to cream or pink to gray

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text