Thermo Scientific Chemicals

(+/-)-Styrene oxide, 98+%, Thermo Scientific Chemicals

Catalog number: L07821.22
100 g, Each
Thermo Scientific Chemicals

(+/-)-Styrene oxide, 98+%, Thermo Scientific Chemicals

Catalog number: L07821.22
100 g, Each
Quantity
Catalog number: L07821.22
also known as L07821-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-09-3
IUPAC Name
2-phenyloxirane
Molecular Formula
C8H8O
InChI Key
AWMVMTVKBNGEAK-UHFFFAOYNA-N
SMILES
C1OC1C1=CC=CC=C1
Form
Liquid
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Refractive Index
1.5330-1.5370 @20°C

Description

Styrene oxide is used as an intermediate in medicine and perfume.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Styrene oxide is used as an intermediate in medicine and perfume.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text