Thermo Scientific Chemicals

3-Aminobenzyl alcohol, 98+%, Thermo Scientific Chemicals

Catalog number: L07847.06
5 g, Each
Thermo Scientific Chemicals

3-Aminobenzyl alcohol, 98+%, Thermo Scientific Chemicals

Catalog number: L07847.06
5 g, Each
Quantity
Catalog number: L07847.06
also known as L07847-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1877-77-6
IUPAC Name
(3-aminophenyl)methanol
Molecular Formula
C7H9NO
InChI Key
OJZQOQNSUZLSMV-UHFFFAOYSA-N
SMILES
NC1=CC=CC(CO)=C1
Melting Point (clear melt)
90.5-96.5?C
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale yellow or cream to brown or grey
Assay (GC)
≥98.0%

Description

3-Aminobenzyl alcohol, is used as a reagent in the synthesis of quinone analogs as dynamin GTPase inhibitors. Also used as a reagent in the synthesis of pyrrolo[2,1-f][1,2,4]triazines as novel hedgehog signaling pathway inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminobenzyl alcohol, is used as a reagent in the synthesis of quinone analogs as dynamin GTPase inhibitors. Also used as a reagent in the synthesis of pyrrolo[2,1-f][1,2,4]triazines as novel hedgehog signaling pathway inhibitors.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text