Thermo Scientific Chemicals

2,6-Dichlorostyrene, 96%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: L07860.14
25 g, Each
Thermo Scientific Chemicals

2,6-Dichlorostyrene, 96%, stab. with 0.1% 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: L07860.14
25 g, Each
Quantity
Catalog number: L07860.14
also known as L07860-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
28469-92-3
Form
Liquid
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.5710-1.5770 @ 20?C
Solution Test
(10% w/v in Methanol): Clear
Assay (GC)
≥95.0%

Description

2,6-Dichlorostyrene is used in the preparation of 1,3-dichloro-2-(1,2-dibromo-ethyl)-benzene by bromination reaction using bromine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichlorostyrene is used in the preparation of 1,3-dichloro-2-(1,2-dibromo-ethyl)-benzene by bromination reaction using bromine.

Solubility
Immiscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text