Thermo Scientific Chemicals

4-Chloro-3-nitrocoumarin, 98%, Thermo Scientific Chemicals

Catalog number: L07908.06
5 g, Each
Thermo Scientific Chemicals

4-Chloro-3-nitrocoumarin, 98%, Thermo Scientific Chemicals

Catalog number: L07908.06
5 g, Each
Quantity
Catalog number: L07908.06
also known as L07908-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38464-20-9
IUPAC Name
4-chloro-3-nitro-2H-chromen-2-one
Molecular Formula
C9H4ClNO4
InChI Key
OFLRQEKOAGDHKT-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=C(Cl)C2=CC=CC=C2OC1=O
Appearance (Color)
Pale yellow to yellow to green
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%

Description

4-Chloro-3-nitrocoumarin, is used in synthesis and antimicrobial activity of new 4-heteroarylamino coumarin derivatives that contain nitrogen and sulfur as heteroatoms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-3-nitrocoumarin, is used in synthesis and antimicrobial activity of new 4-heteroarylamino coumarin derivatives that contain nitrogen and sulfur as heteroatoms.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text