4-Ethylbenzonitrile, 98%, Thermo Scientific Chemicals
4-Ethylbenzonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Ethylbenzonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
10 g
Catalog number L07921.09
also known as L07921-09
Price (USD)/ Each
99.20
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
99.20
Add to cart
4-Ethylbenzonitrile, 98%, Thermo Scientific Chemicals
Catalog numberL07921.09
Price (USD)/ Each
99.20
-
Add to cart
Chemical Identifiers
CAS25309-65-3
IUPAC Name4-ethylbenzonitrile
Molecular FormulaC9H9N
InChI KeySXFFMFAQNAFSLF-UHFFFAOYSA-N
SMILESCCC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5235-1.5275 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only