2-Fluoro-6-iodobenzoic acid, 97%, Thermo Scientific Chemicals
2-Fluoro-6-iodobenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluoro-6-iodobenzoic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L07924.03
also known as L07924-03
Price (USD)/ Each
44.80
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
44.80
Add to cart
2-Fluoro-6-iodobenzoic acid, 97%, Thermo Scientific Chemicals
Catalog numberL07924.03
Price (USD)/ Each
44.80
-
Add to cart
Chemical Identifiers
CAS111771-08-5
IUPAC Name2-fluoro-6-iodobenzoate
Molecular FormulaC7H3FIO2
InChI KeyCYCXAPWOBWWNRK-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=C(F)C=CC=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown
Melting Point (clear melt)121.5-130.5?C
FormPowder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only