1-Chloro-4-ethylbenzene is used in paint & coating industry, rubber industry, water-treatment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Chloro-4-ethylbenzene is used in paint & coating industry, rubber industry, water-treatment.
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Chintada Nageswara Rao.; Shmaryahu Hoz. Synergism and Inhibition in the Combination of Visible Light and HMPA in SmI2 Reductions. J. Org. Chem. 2012, 77, (20), 9199-9204.
Ramesh Yella.; Shmaryahu Hoz. Reduction of 4-Styrylpyridine by SmI2: An Inner Sphere Electron Tranfer Case Where the Binding Site Differs from the Reaction Center. Org. Lett. 2013, 15, (20), 5262-5265.