Thermo Scientific Chemicals

3-(4-Hydroxyphenyl)propionitrile, 98%, Thermo Scientific Chemicals

Catalog number: L07934.06
5 g, Each
Thermo Scientific Chemicals

3-(4-Hydroxyphenyl)propionitrile, 98%, Thermo Scientific Chemicals

Catalog number: L07934.06
5 g, Each
Quantity
Catalog number: L07934.06
also known as L07934-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17362-17-3
IUPAC Name
3-(4-hydroxyphenyl)propanenitrile
Molecular Formula
C9H9NO
InChI Key
KDMJGLYRWRHKJS-UHFFFAOYSA-N
SMILES
OC1=CC=C(CCC#N)C=C1
Form
Crystals or powder or crystalline powder or lumps or fused solid or granules
Appearance (Color)
White to pale cream to pale yellow to pale brown
Assay (GC)
≥97.5%

Description

3-(4-Hydroxyphenyl)propionitrile is commonly used as a tool to evaluate the biological role of ERβ.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Hydroxyphenyl)propionitrile is commonly used as a tool to evaluate the biological role of ERβ.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text