Thermo Scientific Chemicals

Linoleic acid, 95%, Thermo Scientific Chemicals

Catalog number: L07949.22
100 g, Each
Thermo Scientific Chemicals

Linoleic acid, 95%, Thermo Scientific Chemicals

Catalog number: L07949.22
100 g, Each
Quantity
Catalog number: L07949.22
also known as L07949-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60-33-3
IUPAC Name
(9Z,12Z)-octadeca-9,12-dienoic acid
Molecular Formula
C18H32O2
InChI Key
OYHQOLUKZRVURQ-HZJYTTRNSA-N
SMILES
CCCCCC=C/CC=C/CCCCCCCC(O)=O
Appearance (Color)
Clear colorless to yellow to red-yellow
Form
Liquid or viscous liquid
Assay (Silylated GC)
≥94.0% (non-U.S. sourced material)
Assay from Supplier's CofA
≥94.0% (U.S. sourced material)
Comment
Material sourced in the U.S. and in other countries

Description

Unsaturated μ-6 fatty acid Linoleic acid is involved in the production of quick-drying oils which are used in oil paints and varnishes. It is also used in beauty products due to its beneficial properties on the skin such as moisture retention. It is a key starting material for the synthesis of linoleyl alcohol and octadecanoic acid. It is used as a precursor of arachidonic acid (AA) and various prostaglandins. It may be used to improve the delivery and efficacy of anti-cancer drugs and in cancer protection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Unsaturated µ-6 fatty acid Linoleic acid is involved in the production of quick-drying oils which are used in oil paints and varnishes. It is also used in beauty products due to its beneficial properties on the skin such as moisture retention. It is a key starting material for the synthesis of linoleyl alcohol and octadecanoic acid. It is used as a precursor of arachidonic acid (AA) and various prostaglandins. It may be used to improve the delivery and efficacy of anti-cancer drugs and in cancer protection.

Solubility
Miscible with dimethyl sulfoxide, ethanol, ether, petroleum ether, dimethylformamide, acetone and benzene. Insoluble in water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text