Thermo Scientific Chemicals

2-Chlorobenzoxazole, 98%, Thermo Scientific Chemicals

Catalog number: L07981.06
5 g, Each
Thermo Scientific Chemicals

2-Chlorobenzoxazole, 98%, Thermo Scientific Chemicals

Catalog number: L07981.06
5 g, Each
Quantity
Catalog number: L07981.06
also known as L07981-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
615-18-9
IUPAC Name
2-chloro-1,3-benzoxazole
Molecular Formula
C7H4ClNO
InChI Key
BBVQDWDBTWSGHQ-UHFFFAOYSA-N
SMILES
ClC1=NC2=CC=CC=C2O1
Identification (FTIR)
Conforms
Form
Liquid
Refractive Index
1.5650-1.5690 @ 20?C
Appearance (Color)
Clear colorless to yellow to orange
Assay (GC)
≥97.5%

Description

2-Chlorobenzoxazole was used in the synthesis of 2-(2-naphthylamino)benzoxazole via reaction with 2-amino-1-naphthalenesulfonic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzoxazole was used in the synthesis of 2-(2-naphthylamino)benzoxazole via reaction with 2-amino-1-naphthalenesulfonic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Not miscible in water.

Notes
Store under inert gas. Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Keep away from heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text