Thermo Scientific Chemicals

Ethyl 3-(4-methoxybenzoyl)propionate, 98%, Thermo Scientific Chemicals

Catalog number: L07989.14
25 g, Each
Thermo Scientific Chemicals

Ethyl 3-(4-methoxybenzoyl)propionate, 98%, Thermo Scientific Chemicals

Catalog number: L07989.14
25 g, Each
Quantity
Catalog number: L07989.14
also known as L07989-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15118-67-9
IUPAC Name
ethyl 4-(4-methoxyphenyl)-4-oxobutanoate
Molecular Formula
C13H16O4
InChI Key
FYUAOZFEVHSJTO-UHFFFAOYSA-N
SMILES
CCOC(=O)CCC(=O)C1=CC=C(OC)C=C1
Appearance (Color)
White to yellow to cream
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Melting Point (clear melt)
48.0-54.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text