Thermo Scientific Chemicals

5-Methylisatin, 97%, Thermo Scientific Chemicals

Catalog number: L08009.03
1 g, Each
Thermo Scientific Chemicals

5-Methylisatin, 97%, Thermo Scientific Chemicals

Catalog number: L08009.03
1 g, Each
Quantity
Catalog number: L08009.03
also known as L08009-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
608-05-9
IUPAC Name
5-methyl-2,3-dihydro-1H-indole-2,3-dione
Molecular Formula
C9H7NO2
InChI Key
VAJCSPZKMVQIAP-UHFFFAOYSA-N
SMILES
CC1=CC=C2NC(=O)C(=O)C2=C1
Refractive Index
1.4340-1.4390 @ 20?C
Form
Crystals or powder or crystalline powder or lumps or clear liquid as melt
Appearance (Color)
Colorless to dark red to red to dark brown to brown
Assay (HPLC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text