Thermo Scientific Chemicals

Methyl methylsulfonylacetate, 98+%, Thermo Scientific Chemicals

Catalog number: L08039.06
5 g, Each
Thermo Scientific Chemicals

Methyl methylsulfonylacetate, 98+%, Thermo Scientific Chemicals

Catalog number: L08039.06
5 g, Each
Quantity
Catalog number: L08039.06
also known as L08039-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62020-09-1
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥98.0%
Appearance (Color)
White to pale cream to pale yellow

Description

Methyl methylsulfonylacetate is used as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl methylsulfonylacetate is used as an active pharmaceutical ingredient.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text