Methyl methylsulfonylacetate, 98+%, Thermo Scientific Chemicals
Methyl methylsulfonylacetate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl methylsulfonylacetate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L08039.14
also known as L08039-14
Price (USD)/ Each
314.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
314.00
Add to cart
Methyl methylsulfonylacetate, 98+%, Thermo Scientific Chemicals
Catalog numberL08039.14
Price (USD)/ Each
314.00
-
Add to cart
Chemical Identifiers
CAS62020-09-1
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥98.0%
Appearance (Color)White to pale cream to pale yellow
Methyl methylsulfonylacetate is used as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl methylsulfonylacetate is used as an active pharmaceutical ingredient.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Barry M. Trost.; Paul E. Strege. Regio- and stereoselectivity of allylic alkylation. J. Am. Chem. Soc. 1975, 97, (9), 2534-2535.
  2. Barry M. Trost.; Thomas J. Dietsche.; Terry J. Fullerton. New synthetic reactions. Chemospecificity of allylic alkylation. J. Org. Chem. 1974, 39, (5), 737-738.