Thermo Scientific Chemicals

2-Methoxy-3-methylpyrazine, 99%, Thermo Scientific Chemicals

Catalog number: L08067.06
5 g, Each
Thermo Scientific Chemicals

2-Methoxy-3-methylpyrazine, 99%, Thermo Scientific Chemicals

Catalog number: L08067.06
5 g, Each
Quantity
Catalog number: L08067.06
also known as L08067-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2847-30-5
IUPAC Name
2-methoxy-3-methylpyrazine
Molecular Formula
C6H8N2O
InChI Key
VKJIAEQRKBQLLA-UHFFFAOYSA-N
SMILES
COC1=NC=CN=C1C
Refractive Index
1.5060-1.5090 @ 20?C
Form
Liquid
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless to pale yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text