2-Methoxy-3-methylpyrazine, 99%, Thermo Scientific Chemicals
2-Methoxy-3-methylpyrazine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methoxy-3-methylpyrazine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number L08067.14
also known as L08067-14
Price (USD)/ Each
185.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
185.00
Add to cart
2-Methoxy-3-methylpyrazine, 99%, Thermo Scientific Chemicals
Catalog numberL08067.14
Price (USD)/ Each
185.00
-
Add to cart
Chemical Identifiers
CAS2847-30-5
IUPAC Name2-methoxy-3-methylpyrazine
Molecular FormulaC6H8N2O
InChI KeyVKJIAEQRKBQLLA-UHFFFAOYSA-N
SMILESCOC1=NC=CN=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5060-1.5090 @ 20?C
FormLiquid
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only