Thermo Scientific Chemicals

L-Noradrenaline, 98%, Thermo Scientific Chemicals

Catalog number: L08087.03
1 g, Each
Thermo Scientific Chemicals

L-Noradrenaline, 98%, Thermo Scientific Chemicals

Catalog number: L08087.03
1 g, Each
Quantity
Catalog number: L08087.03
also known as L08087-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51-41-2
IUPAC Name
4-[(1R)-2-amino-1-hydroxyethyl]benzene-1,2-diol
Molecular Formula
C8H11NO3
InChI Key
SFLSHLFXELFNJZ-QMMMGPOBSA-N
SMILES
NC[C@H](O)C1=CC=C(O)C(O)=C1
Assay (HPLC)
≥97.5%
Optical Rotation
-47.2 ± 0.5° (C=5 in 1M HCl)
Form
Powder
Identification (FTIR)
Conforms (non-U.S. specification)
Appearance (Color)
White to cream to brown to pale gray

Description

L-Noradrenaline serves as an adrenergic neurotransmitter. It acts as demethylated precursor of epinephrine (adrenaline). It is also used to treat people having low blood pressure, as a sympathomimetic drug to treat people in vasodilatory shock states such as septic shock and neurogenic shock. Further, it is used in the preparation of1,2,3,4-tetrahydro-4,6,7-isoquinolinetriol hydrochloride. In addition to this, it is used in the treatment of hypotension, depression, schizophrenia and attention hyperactivity disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Noradrenaline serves as an adrenergic neurotransmitter. It acts as demethylated precursor of epinephrine (adrenaline). It is also used to treat people having low blood pressure, as a sympathomimetic drug to treat people in vasodil
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text