Thermo Scientific Chemicals

trans-2-Methyl-2-butenal, 97%, Thermo Scientific Chemicals

Catalog number: L08113.14
25 g, Each
Thermo Scientific Chemicals

trans-2-Methyl-2-butenal, 97%, Thermo Scientific Chemicals

Catalog number: L08113.14
25 g, Each
Quantity
Catalog number: L08113.14
also known as L08113-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
497-03-0
IUPAC Name
(2E)-2-methylbut-2-enal
Molecular Formula
C5H8O
InChI Key
ACWQBUSCFPJUPN-HWKANZROSA-N
SMILES
C\C=C(/C)C=O
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.4455-1.4505 @ 20?C
Free acid (titration)
≤3.0%
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms

Description

Tiglic aldehyde was used in the concise total synthesis of 7-demethylpiericidin A11. It was used as starting reagent during the synthesis of alkyl-branched tetraene hydrocarbons, pheromone component for dried fruit beetle.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tiglic aldehyde was used in the concise total synthesis of 7-demethylpiericidin A11. It was used as starting reagent during the synthesis of alkyl-branched tetraene hydrocarbons, pheromone component for dried fruit beetle.

Solubility
Soluble in water

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text