Thermo Scientific Chemicals

Magnesium, turnings, 99+%, Thermo Scientific Chemicals

Catalog number: L08120.0E
2500 g, Each
Thermo Scientific Chemicals

Magnesium, turnings, 99+%, Thermo Scientific Chemicals

Catalog number: L08120.0E
2500 g, Each
Quantity
Catalog number: L08120.0E
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7439-95-4
IUPAC Name
magnesium
Molecular Formula
Mg
InChI Key
JLVVSXFLKOJNIY-UHFFFAOYSA-N
SMILES
[Mg]
Assay (unspecified)
> 99.0% with respect to metal impurities
Form
metal raspings
Appearance (Color)
Shiny, silver

Description

Magnesium, turnings is used in solid state synthesis with Ca and Sn resulted in a new phase, Ca6.2Mg3.8Sn7, which has an unprecedented type of tin chain composed of square-planar tin units. It is a component in die-casting, to remove sulfur in the production of iron and steel, the production of titanium in the Kroll process. It is widely used for manufacturing of mobile phones, laptop and tablet computers, cameras, and other electronic components.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium, turnings is used in solid state synthesis with Ca and Sn resulted in a new phase, Ca6.2Mg3.8Sn7, which has an unprecedented type of tin chain composed of square-planar tin units. It is a component in die-casting, to remove sulfur in the production of iron and steel, the production of titanium in the Kroll process. It is widely used for manufacturing of mobile phones, laptop and tablet computers, cameras, and other electronic components.

Solubility
Reacts with water.

Notes
Store away from air, moisture and oxidizing agents. Incompatible with acids, air, nitrogen and moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text