Thermo Scientific Chemicals

Triphenyl phosphate, 98%, Thermo Scientific Chemicals

Catalog number: L08130.0B
1000 g, Each
Thermo Scientific Chemicals

Triphenyl phosphate, 98%, Thermo Scientific Chemicals

Catalog number: L08130.0B
1000 g, Each
Quantity
Catalog number: L08130.0B
also known as L08130-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
115-86-6
IUPAC Name
triphenyl phosphate
Molecular Formula
C18H15O4P
InChI Key
XZZNDPSIHUTMOC-UHFFFAOYSA-N
SMILES
O=P(OC1=CC=CC=C1)(OC1=CC=CC=C1)OC1=CC=CC=C1
Appearance (Color)
White
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or fused solid or flakes or pellets

Description

Triphenyl phosphate is used as an internal standard in the screening and quantification of agrochemical residues in vegetables and fruits. It is used in plastic processing and molding of plastic flow and performance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenyl phosphate is used as an internal standard in the screening and quantification of agrochemical residues in vegetables and fruits. It is used in plastic processing and molding of plastic flow and performance.

Solubility
Reacts with water.

Notes
Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text