1,2,3,4-Tetrahydroisoquinoline, 97%, Thermo Scientific Chemicals
1,2,3,4-Tetrahydroisoquinoline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2,3,4-Tetrahydroisoquinoline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number L08143.22
also known as L08143-22
Price (USD)/ Each
133.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
133.00
Add to cart
1,2,3,4-Tetrahydroisoquinoline, 97%, Thermo Scientific Chemicals
Catalog numberL08143.22
Price (USD)/ Each
133.00
-
Add to cart
Chemical Identifiers
CAS91-21-4
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5655-1.5710 @ 20°C
Appearance (Color)Clear colorless to yellow
FormLiquid or viscous liquid
Assay (GC)≥95.0%
It is used as a reagent in the preparation of4-(1,2,4-oxadiazol-5-yl)piperidine-1-carboxamides as antiproliferative tubulin inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent in the preparation of4-(1,2,4-oxadiazol-5-yl)piperidine-1-carboxamides as antiproliferative tubulin inhibitors.

Solubility
Soluble in water at 20°C 20g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.
RUO – Research Use Only

General References:

  1. Hiroshi Kawai, et al. Novel Endogenous 1,2,3 ,4-Tetrahydroisoquinoline Derivatives: Uptake by Dopamine Transporter and Activity to Induce Parkinsonism.J. Neurochem.,1998,70(2), 745-751.
  2. Estefanía Méndez-Álvarez, et al. Inhibition of brain monoamine oxidase by adducts of 1,2,3,4-tetrahydroisoquinoline with components of cigarette smoke.Life Sci.,1997,60(19), 1719-1727.