Thermo Scientific Chemicals

Tetra-n-dodecylammonium iodide, 98%, Thermo Scientific Chemicals

Catalog number: L08153.09
10 g, Each
Thermo Scientific Chemicals

Tetra-n-dodecylammonium iodide, 98%, Thermo Scientific Chemicals

Catalog number: L08153.09
10 g, Each
Quantity
Catalog number: L08153.09
also known as L08153-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
40797-39-5
IUPAC Name
tetradodecylazanium iodide
Molecular Formula
C48H100IN
InChI Key
OBMLOTDECOTAGU-UHFFFAOYSA-M
SMILES
[I-].CCCCCCCCCCCC[N+](CCCCCCCCCCCC)(CCCCCCCCCCCC)CCCCCCCCCCCC
Elemental Analysis
Carbon, C: 68.70 - 72.26% (Theory 70.46%)
Elemental Analysis
Iodine, I: 15.12 - 15.90% (Theory 15.51%)
Form
Powder
Elemental Analysis
Hydrogen, H: 12.01 - 12.63% (Theory 12.32%)
Elemental Analysis
Nitrogen, N: 1.67 - 1.75%(Theory 1.71%)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text