Thermo Scientific Chemicals

2,2,2-Trichloroethanol, 99%, Thermo Scientific Chemicals

Catalog number: L08163.14
25 g, Each
Thermo Scientific Chemicals

2,2,2-Trichloroethanol, 99%, Thermo Scientific Chemicals

Catalog number: L08163.14
25 g, Each
Quantity
Catalog number: L08163.14
also known as L08163-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
115-20-8
IUPAC Name
2,2,2-trichloroethan-1-ol
Molecular Formula
C2H3Cl3O
InChI Key
KPWDGTGXUYRARH-UHFFFAOYSA-N
SMILES
OCC(Cl)(Cl)Cl
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.4880-1.4910 @ 20?C
Form
Liquid

Description

2,2,2-Trichloroethanol is used as a sedative or a hypnotic. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trichloroethanol is used as a sedative or a hypnotic. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with oxidizing agents, Strong acids, Strong bases, Strong reducing agents, Acid chlorides, Acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text