Thermo Scientific Chemicals

2-Butanone oxime, 99%, Thermo Scientific Chemicals

Catalog number: L08196.AU
1000 mL, Each
Thermo Scientific Chemicals

2-Butanone oxime, 99%, Thermo Scientific Chemicals

Catalog number: L08196.AU
1000 mL, Each
Quantity
Catalog number: L08196.AU
also known as L08196-AU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-29-7
IUPAC Name
(E)-N-(butan-2-ylidene)hydroxylamine
Molecular Formula
C4H9NO
InChI Key
WHIVNJATOVLWBW-SNAWJCMRSA-N
SMILES
CC\C(C)=N\O
Form
Liquid or viscous liquid
Assay (GC)
≥98.5%
Refractive Index
1.4425-1.4455 @ 20?C
Appearance (Color)
Clear colorless

Description

2-Butanone oxime was employed as reagent and solvent in the syntheses of ketoimine and 2,4-dipyridyl-1,3,5-triazapentadiene palladium(II) complexes. It was also used in the synthesis of novel acetaldiimine cobalt complex, [CoI2{((CH3CH2)(CH3)C=NO)2C(CH3)2}]. Methyl ethyl ketoxime is used primarily as an antiskinning agent in alkyd coating resins

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butanone oxime was employed as reagent and solvent in the syntheses of ketoimine and 2,4-dipyridyl-1,3,5-triazapentadiene palladium(II) complexes. It was also used in the synthesis of novel acetaldiimine cobalt complex, [CoI2{((CH3CH2)(CH3)C=NO)2C(CH3)2}]. Methyl ethyl ketoxime is used primarily as an antiskinning agent in alkyd coating resins

Solubility
Soluble in water at 20°C 114 g/L. Soluble in most organic solvents .

Notes
Store away from acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text