Thermo Scientific Chemicals

4,4'-Dibromobenzophenone, 98+%, Thermo Scientific Chemicals

Catalog number: L08200.03
1 g, Each
Thermo Scientific Chemicals

4,4'-Dibromobenzophenone, 98+%, Thermo Scientific Chemicals

Catalog number: L08200.03
1 g, Each
Quantity
Catalog number: L08200.03
also known as L08200-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3988-03-2
IUPAC Name
bis(4-bromophenyl)methanone
Molecular Formula
C13H8Br2O
InChI Key
LFABNOYDEODDFX-UHFFFAOYSA-N
SMILES
BrC1=CC=C(C=C1)C(=O)C1=CC=C(Br)C=C1
Appearance (Color)
White to cream
Assay (GC)
≥98.0%
Melting Point (clear melt)
172-178?C
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

4,4'-Dibromobenzophenone is used in the synthesis of high molecular weight poly(p-phenylene) derivatives through palladium-catalyzed polycondensation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dibromobenzophenone is used in the synthesis of high molecular weight poly(p-phenylene) derivatives through palladium-catalyzed polycondensation reaction.

Solubility
Soluble in ethanol, 2-propanol, acetone and toluene. Miscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text