2-Chloro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
2-Chloro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L08207.14
also known as L08207-14
Price (USD)/ Each
65.30
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
65.30
Add to cart
2-Chloro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
Catalog numberL08207.14
Price (USD)/ Each
65.30
-
Add to cart
Chemical Identifiers
CAS52334-81-3
IUPAC Name2-chloro-5-(trifluoromethyl)pyridine
Molecular FormulaC6H3ClF3N
InChI KeyJFZJMSDDOOAOIV-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(Cl)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to pale cream to cream to yellow to green
FormCrystals or powder or crystalline powder or lumps or chunks or fused solid or clear liquid as melt
Assay (GC)≥96.0%
Melting Point (clear melt)26.0-35.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only