Thermo Scientific Chemicals

5-Methoxypiperonal, 97%, Thermo Scientific Chemicals

Catalog number: L08213.14
25 g, Each
Thermo Scientific Chemicals

5-Methoxypiperonal, 97%, Thermo Scientific Chemicals

Catalog number: L08213.14
25 g, Each
Quantity
Catalog number: L08213.14
also known as L08213-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5780-07-4
IUPAC Name
7-methoxy-2H-1,3-benzodioxole-5-carbaldehyde
Molecular Formula
C9H8O4
InChI Key
LOFRBHZYZCIOJO-UHFFFAOYSA-N
SMILES
COC1=CC(C=O)=CC2=C1OCO2
Assay (GC)
≥96.0%
Free acid (titration)
≤1.5%
Form
Powder
Appearance (Color)
White to cream to brown or grey
Melting Point (clear melt)
129.0-135.0?C

Description

5-Methoxypiperonal is used in the synthesis of oxazole combretastatin A-4 analogs showing anti-tumor activity. It is also used in the synthesis of endothelin-A antagonist ABT-546, and further in the synthesis of antidepressant rolipram.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methoxypiperonal is used in the synthesis of oxazole combretastatin A-4 analogs showing anti-tumor activity. It is also used in the synthesis of endothelin-A antagonist ABT-546, and further in the synthesis of antidepressant rolipram.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with air, oxidizing agents. Store under dry inert gas. Store away from air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text