tert-Butyl 2-bromoisobutyrate, 97%, Thermo Scientific Chemicals
tert-Butyl 2-bromoisobutyrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyl 2-bromoisobutyrate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number L08216.30
also known as L08216-30
Price (USD)/ Each
219.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
219.00
Add to cart
tert-Butyl 2-bromoisobutyrate, 97%, Thermo Scientific Chemicals
Catalog numberL08216.30
Price (USD)/ Each
219.00
-
Add to cart
Chemical Identifiers
CAS23877-12-5
IUPAC Nametert-butyl 2-bromo-2-methylpropanoate
Molecular FormulaC8H15BrO2
InChI KeyIGVNJALYNQVQIT-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)C(C)(C)Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4340-1.4390 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only