4-Nitro-3-(trifluoromethyl)phenol, 97%, Thermo Scientific Chemicals
4-Nitro-3-(trifluoromethyl)phenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Nitro-3-(trifluoromethyl)phenol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L08228.03
also known as L08228-03
Price (USD)/ Each
35.60
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
35.60
Add to cart
4-Nitro-3-(trifluoromethyl)phenol, 97%, Thermo Scientific Chemicals
Catalog numberL08228.03
Price (USD)/ Each
35.60
-
Add to cart
Chemical Identifiers
CAS88-30-2
IUPAC Name4-nitro-3-(trifluoromethyl)phenol
Molecular FormulaC7H4F3NO3
InChI KeyZEFMBAFMCSYJOO-UHFFFAOYSA-N
SMILESOC1=CC=C(C(=C1)C(F)(F)F)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow
FormCrystals or crystalline powder or powder
Assay (GC)≥96.0%
Melting Point (clear melt)71-80°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only