D-(-)-Norleucine, 99%
D-(-)-Norleucine, 99%
Thermo Scientific Chemicals

D-(-)-Norleucine, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L08257.03
also known as L08257-03
Price (USD)/ Each
56.80
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
56.80
Add to cart
D-(-)-Norleucine, 99%
Catalog numberL08257.03
Price (USD)/ Each
56.80
-
Add to cart
Chemical Identifiers
CAS327-56-0
IUPAC Name(2R)-2-aminohexanoic acid
Molecular FormulaC6H13NO2
InChI KeyLRQKBLKVPFOOQJ-RXMQYKEDSA-N
SMILESCCCC[C@@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or flakes
Optical Rotation-23.3 ± 0.1? (C=5 in 5N HCl)
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only