N-Benzoylaminopurine, 99%, Thermo Scientific Chemicals
N-Benzoylaminopurine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzoylaminopurine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number L08292.06
also known as L08292-06
Price (USD)/ Each
188.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
188.00
Add to cart
N-Benzoylaminopurine, 99%, Thermo Scientific Chemicals
Catalog numberL08292.06
Price (USD)/ Each
188.00
-
Add to cart
Chemical Identifiers
CAS4005-49-6
IUPAC NameN-(7H-purin-6-yl)benzamide
Molecular FormulaC12H9N5O
InChI KeyQQJXZVKXNSFHRI-UHFFFAOYSA-N
SMILESO=C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Appearance (Color)White to cream to pale brown
Assay (HPLC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only