Thermo Scientific Chemicals

(Phenylsulfonyl)acetic acid, 97%, Thermo Scientific Chemicals

Catalog number: L08299.14
25 g, Each
Thermo Scientific Chemicals

(Phenylsulfonyl)acetic acid, 97%, Thermo Scientific Chemicals

Catalog number: L08299.14
25 g, Each
Quantity
Catalog number: L08299.14
also known as L08299-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3959-23-7
IUPAC Name
2-(benzenesulfonyl)acetic acid
Molecular Formula
C8H8O4S
InChI Key
YTEFAALYDTWTLB-UHFFFAOYSA-N
SMILES
OC(=O)CS(=O)(=O)C1=CC=CC=C1
Appearance (Color)
White to grey to pale brown or pale yellow
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
108.0-115.0°C
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

(Phenylsulfonyl)acetic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Phenylsulfonyl)acetic acid is used as pharmaceutical intermediates.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text