Thermo Scientific Chemicals

(R)-(+)-1,1'-Bi(2-naphthol), 99%, Thermo Scientific Chemicals

Catalog number: L08305.03
1 g, Each
Thermo Scientific Chemicals

(R)-(+)-1,1'-Bi(2-naphthol), 99%, Thermo Scientific Chemicals

Catalog number: L08305.03
1 g, Each
Quantity
Catalog number: L08305.03
also known as L08305-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18531-94-7
IUPAC Name
[1,1'-binaphthalene]-2,2'-diol
Molecular Formula
C20H14O2
InChI Key
PPTXVXKCQZKFBN-UHFFFAOYSA-N
SMILES
OC1=CC=C2C=CC=CC2=C1C1=C(O)C=CC2=CC=CC=C12
Appearance (Color)
White to cream or pale gray
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
205-212°C
Optical Rotation
+35.5° ± 2° (c=1 in THF)
Assay (HPLC)
≥98.5%

Description

It is used in biosynthetic preparation for enantioselective oxidation of naphthols to binaphthyldiols with horseradish peroxidase catalyst. A chiral auxiliary used in the catalytic asymmetric oxidation of sulfides to sulfoxides. Chiral lanthanide triflates formed from binaphthol serve as catalysts for asymmetric Diels-Alder reactions. Derivatives of binaphthol have recently found use in asymmetric Claisen rearrangements and asymmetric epoxidations.5 The lithium aluminum hydride derivative of these diols (BINAP-H) has been used extensively for the reduction of ketones. Chiral binapthol imminium salt precursor. Salts were used for an asymmetric epoxidation of olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in biosynthetic preparation for enantioselective oxidation of naphthols to binaphthyldiols with horseradish peroxidase catalyst. A chiral auxiliary used in the catalytic asymmetric oxidation of sulfides to sulfoxides. Chiral lanthanide triflates formed from binaphthol serve as catalysts for asymmetric Diels-Alder reactions. Derivatives of binaphthol have recently found use in asymmetric Claisen rearrangements and asymmetric epoxidations.5 The lithium aluminum hydride derivative of these diols (BINAP-H) has been used extensively for the reduction of ketones. Chiral binapthol imminium salt precursor. Salts were used for an asymmetric epoxidation of olefins.

Solubility
Insoluble in water. Solubility in THF within almost transparency). Soluble in dioxane 50 mg/mL.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text