3,5-Dichloro-2-pyridone, 98+%, Thermo Scientific Chemicals
3,5-Dichloro-2-pyridone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dichloro-2-pyridone, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L08309.03
also known as L08309-03
Price (USD)/ Each
63.80
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
63.80
Add to cart
3,5-Dichloro-2-pyridone, 98+%, Thermo Scientific Chemicals
Catalog numberL08309.03
Price (USD)/ Each
63.80
-
Add to cart
Chemical Identifiers
CAS5437-33-2
IUPAC Name3,5-dichloro-1,2-dihydropyridin-2-one
Molecular FormulaC5H3Cl2NO
InChI KeyZICOPWJJZSJEDL-UHFFFAOYSA-N
SMILESClC1=CNC(=O)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Melting Point (clear melt)178-184?C
Appearance (Color)White to cream to grey
Assay (HPLC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only